Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR37 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4450
This item is not returnable.
View return policy
Description
GPR37 Monoclonal specifically detects GPR37 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
GPR37 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
O15354 | |
GPR37 | |
NS0 mouse myeloma cell line transfected with human GPR37 Ala27-Cys613 Accession # O15354 | |
100 μg | |
Primary | |
Detects human GPR37. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
436923 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EDNRBL, Endothelin B receptor-like protein 1, ETBR-LP-1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELR, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2861 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction