Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR39 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3348V100UG
This item is not returnable.
View return policy
Description
GPR39 Monoclonal specifically detects GPR39 in Human samples. It is validated for Flow Cytometry.Specifications
| GPR39 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| BAG37409 | |
| GPR39 | |
| NS0 mouse myeloma cell line transfected with human GPR39 Met1-Val453 Accession # BAG37409 | |
| RUO | |
| 2863 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 189702 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GPR39. Stains human GPR39-transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction