Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR50 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Catalog No. IC4645T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4645T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4645T100 Supplier R&D Systems Supplier No. IC4645T100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR50 Monoclonal specifically detects GPR50 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen GPR50
Applications Flow Cytometry
Classification Monoclonal
Clone 461129
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q13585
Gene Alias G protein-coupled receptor 50H9Mel1c, H9, melatonin-related receptor, MGC125342, MTR1L
Gene Symbols GPR50
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR50 Met1-Val617 Accession # Q13585
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9248
Test Specificity Detects human GPR50. Stains human GPR50 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.