Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR56 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB46361T100UG
This item is not returnable.
View return policy
Description
GPR56 Monoclonal specifically detects GPR56 in Human samples. It is validated for Flow Cytometry.Specifications
GPR56 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
BFPP, EGF-TM7-like, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4, TM7LN4G-protein coupled receptor 56, TM7XN1, TM7XN1DKFZp781L1398 | |
Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val345, Accession # Q9Y653 | |
100 μg | |
Primary | |
Detects human GPR56 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
2446B | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y653 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
9289 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction