Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR56 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4636R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4636R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4636R100 Supplier R&D Systems Supplier No. FAB4636R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR56 Monoclonal specifically detects GPR56 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GPR56
Applications Flow Cytometry
Classification Monoclonal
Clone 776123
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9Y653
Gene Alias BFPP, EGF-TM7-like, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4, TM7LN4G-protein coupled receptor 56, TM7XN1, TM7XN1DKFZp781L1398
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val342, Accession No. Q9Y653
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9289.0
Test Specificity Detects human GPR56 in direct ELISA.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.