Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR64 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB79771T100UG
This item is not returnable.
View return policy
Description
GPR64 Monoclonal specifically detects GPR64 in Human samples. It is validated for Flow Cytometry.Specifications
GPR64 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, HE6, MGC104454, MGC138738, TM7LN2 | |
Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession # NP_001073328 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
864238 | |
Flow Cytometry 0.25-1 ug/106 cells | |
NP_001073328 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10149 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction