Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR75 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.00
Specifications
Antigen | GPR75 |
---|---|
Clone | 689714 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB67281SP
![]() |
R&D Systems
MAB67281SP |
25 μg |
Each for $200.50
|
|
|||||
MAB67281100
![]() |
R&D Systems
MAB67281100 |
100 μg |
Each for $444.00
|
|
|||||
Description
GPR75 Monoclonal antibody specifically detects GPR75 in Human samples. It is validated for Flow CytometrySpecifications
GPR75 | |
Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
G protein-coupled receptor 75, GPRchr2, MGC132002, probable G-protein coupled receptor 75, WI31133, WI-31133 | |
NS0 mouse myeloma cell line transfected with human GPR75, Accession # O95800 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
689714 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
O95800 | |
10936 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title