Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR81 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | GPR81 |
---|---|
Clone | 1048103 |
Dilution | Western Blot 0.5 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11134SP
![]() |
R&D Systems
MAB11134SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11134100
![]() |
R&D Systems
MAB11134100 |
100 μg |
Each for $502.50
|
|
|||||
Description
GPR81 Monoclonal antibody specifically detects GPR81 in Human samples. It is validated for Western BlotSpecifications
GPR81 | |
Western Blot 0.5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
27198 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1048103 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
FKSG80, G protein-coupled receptor 104, G protein-coupled receptor 81, GPR104, G-protein coupled receptor 104, G-protein coupled receptor 81, HCA1, HCAR1, LACR1, lactate receptor 1, TA-GPCR, T-cell activation G protein-coupled receptor | |
Synthetic peptide corresponding to the N-term sequences of Human HCAR1/GPR81, | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title