Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPR97 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB10097S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB10097S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB10097S100 Supplier R&D Systems Supplier No. FAB10097S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GPR97 Monoclonal specifically detects GPR97 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GPR97
Applications Flow Cytometry
Classification Monoclonal
Clone 2356C
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Gene Accession No. Q86Y34
Gene Alias G protein-coupled receptor 97, Pb99, PGR26, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession No. Q86Y34
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 222487.0
Test Specificity Detects human GPR97 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.