Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR97 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $467.50
Specifications
Antigen | GPR97 |
---|---|
Clone | 2435C |
Dilution | Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10097SP
![]() |
R&D Systems
MAB10097SP |
25 μg |
Each for $214.00
|
|
|||||
MAB10097100
![]() |
R&D Systems
MAB10097100 |
100 μg |
Each for $467.50
|
|
|||||
Description
GPR97 Monoclonal specifically detects GPR97 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
GPR97 | |
Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
Q86Y34 | |
222487 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2435C | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
G protein-coupled receptor 97, Pb99, PGR26, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97 | |
Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession # Q86Y34 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GPR97 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title