Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPRC5B Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10253S10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10253S10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10253S10 Supplier R&D Systems Supplier No. FAB10253S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPRC5B Monoclonal specifically detects GPRC5B in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GPRC5B
Applications Flow Cytometry
Classification Monoclonal
Clone 575926
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9NZH0
Gene Alias G protein-coupled receptor, family C, group 1, member B, G protein-coupled receptor, family C, group 5, member B, RAIG2, RAIG-2A-69G12.1, RAIG2G-protein coupled receptor family C group 5 member B, retinoic acid responsive gene protein, Retinoic acid-induced gene 2 protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPRC5B, Accession # Q9NZH0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51704
Test Specificity Detects human GPRC5B in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.