Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GPRC5C Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6594U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6594U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6594U100 Supplier R&D Systems Supplier No. FAB6594U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPRC5C Monoclonal specifically detects GPRC5C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GPRC5C
Applications Flow Cytometry
Classification Monoclonal
Clone 577315
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9NQ84
Gene Alias G protein-coupled receptor, family C, group 5, member C, MGC131820, RAIG3, RAIG-3G-protein coupled receptor family C group 5 member C, RAIG3orphan G-protein coupled receptor, retinoic acid responsive gene protein, Retinoic acid-induced gene 3 protein
Gene Symbols GPRC5C
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with recombinant human GPRC5C Accession # Q9NQ84
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55890
Test Specificity Detects human GPRC5C in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.