Learn More
Human GPRC5D Alexa Fluor 647-conjugated Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | GPRC5D |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 571961R |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q9NZD1 |
| Gene Alias | G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.