Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPRC5D PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6300RP100
This item is not returnable.
View return policy
Description
GPRC5D Monoclonal specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry.Specifications
GPRC5D | |
Monoclonal | |
PE | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
NS0 mouse myeloma cell line transfected with human GPRC5D, Met1-Val359, Accession # Q9NZD1 | |
100 Tests | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
571961R | |
Flow Cytometry 10 uL/10^6 cells | |
Q9NZD1 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
55507 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction