Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Granulysin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | Granulysin |
---|---|
Clone | 786314 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB31381SP
![]() |
R&D Systems
MAB31381SP |
25 μg |
Each for $200.50
|
|
|||||
MAB31381100
![]() |
R&D Systems
MAB31381100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISASpecifications
Granulysin | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
D2S69Elymphocyte-activation gene 2, GNLY, granulysin, LAG2, LAG-2, Lymphokine, Lymphokine LAG-2, NKG5, NKG5519, Protein NKG5, TLA519, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519 | |
Mouse myeloma cell line NS0-derived recombinant human Granulysin, Arg23-Leu145, Accession # P22749 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
786314 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P22749 | |
10578 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title