Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Granzyme B Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2906G
This item is not returnable.
View return policy
Description
Granzyme B Monoclonal specifically detects Granzyme B in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Granzyme B | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3002 | |
Human | |
Purified |
Flow Cytometry | |
351927 | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
P10144 | |
GZMB | |
Mouse myeloma cell line NS0-derived recombinant human Granzyme B Gly19-Tyr247 Accession # P10144 | |
100 Tests | |
Primary | |
Detects human Granzyme B in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Granzyme A, rhGranzyme H, recombinant mouse (rm) Granzyme B, rmGranzyme C, rmGranzyme D, or rmGranzyme G. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction