Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Granzyme B PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC2906C
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2906C 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2906C Supplier R&D Systems Supplier No. IC2906C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Granzyme B Monoclonal specifically detects Granzyme B in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Granzyme B
Applications Flow Cytometry
Classification Monoclonal
Clone 351927
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P10144
Gene Alias C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT
Gene Symbols GZMB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Granzyme B Gly19-Tyr247 Accession # P10144
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3002
Test Specificity Detects human Granzyme B in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Granzyme A, rhGranzyme H, recombinant mouse (rm) Granzyme B, rmGranzyme C, rmGranzyme D, or rmGranzyme G.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.