Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Granzyme H Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1377SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1377SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1377SP Supplier R&D Systems Supplier No. AF1377SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Granzyme H Polyclonal specifically detects Granzyme H in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Granzyme H
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20718
Gene Alias Cathepsin G-like 2, CCP-X, CCP-Xcytotoxic T-lymphocyte-associated serine esterase 1, CGL2, CGL-2, CSP-Ccytotoxin serine protease-C, CTLA1, CTSGL2, CTSGL2granzyme H, Cytotoxic serine protease C, Cytotoxic T-lymphocyte proteinase, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.79, granzyme H (cathepsin G-like 2, protein h-CCPX), GrzH, GZMH
Gene Symbols GZMH
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Granzyme H Glu19-Leu246 Accession # P20718
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2999
Test Specificity Detects both the pro and active forms of human Granzyme H in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human Granzyme B is observed and approximately 5% cross-reactivity with recombinant mouse (rm) Granzyme C, rmGranzyme D, and rmGranzyme G is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.