Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Granzyme H Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Granzyme H |
---|---|
Clone | 185813 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Granzyme H Monoclonal specifically detects Granzyme H in Human samples. It is validated for Western Blot.Specifications
Granzyme H | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P20718 | |
2999 | |
Mouse myeloma cell line NS0-derived recombinant human Granzyme H Glu19-Leu246 Accession # P20718 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Granzyme H in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Granzyme D or rmGranzyme G is observed. |
185813 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cathepsin G-like 2, CCP-X, CCP-Xcytotoxic T-lymphocyte-associated serine esterase 1, CGL2, CGL-2, CSP-Ccytotoxin serine protease-C, CTLA1, CTSGL2, CTSGL2granzyme H, Cytotoxic serine protease C, Cytotoxic T-lymphocyte proteinase, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.79, granzyme H (cathepsin G-like 2, protein h-CCPX), GrzH, GZMH | |
GZMH | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title