Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Granzyme K Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $502.50
Specifications
Antigen | Granzyme K |
---|---|
Clone | 2471A |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 25 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10216SP
![]() |
R&D Systems
MAB10216SP |
25 μg |
Each for $214.00
|
|
|||||
MAB10216100
![]() |
R&D Systems
MAB10216100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Granzyme K Monoclonal specifically detects Granzyme K in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Granzyme K | |
Western Blot 0.1 ug/mL, Flow Cytometry 25 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P49863 | |
3003 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2471A | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, Granzyme 3, granzyme K, granzyme K (granzyme 3; tryptase II), granzyme K (serine protease, granzyme 3; tryptase II), Granzyme-3, Granzyme-K, GZMK, NK-Tryp-2, NK-Tryptase-2, PRSS, TRYP2, TRYP2granzyme-3, tryptase II | |
Synthetic peptide containing human Granzyme K, Accession # P49863 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Granzyme K in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title