Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Granzyme K ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Assay Range | 31.25 - 2000 pg/mL |
Assay Sensitivity | 18.75 pg/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Biological Fluids, Plasma, Serum |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 3003 |
Gene Symbol | GZMK |
Quantity | 1 Kit |
Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Synonym | EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, granzyme 3, granzyme K, granzyme K (granzyme 3; tryptase II), granzyme K (serine protease, granzyme 3; tryptase II), Granzyme-3, NK-Tryp-2, NK-tryptase-2, PRSS, TRYP2granzyme-3, tryptase II |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction