Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRB14 Partial ORF (NP_004481, 25 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7094421
Click to view available options
Quantity:
10 ug
25 ug

Used for AP, Array, ELISA, WB-Re

The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. This protein likely has an inhibitory effect on receptor tyrosine kinase signaling and, in particular, on insulin receptor signaling. This gene may play a role in signaling pathways that regulate growth and metabolism. Transcript variants have been reported for this gene, but their full-length natures have not been determined to date. [provided by RefSeq]

Sequence: QVCGAAQGRGDAHDLAPAPWLHARALLPLPDGTRGCAADRRKKKDLDVPEMPSIPNPFPELCCSPITSVLSADLFPKANSRKKQVIKVYSEDETSRALDV

Specifications

Accession Number NP_004481
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2888
Molecular Weight (g/mol) 36.74kDa
Name GRB14 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen QVCGAAQGRGDAHDLAPAPWLHARALLPLPDGTRGCAADRRKKKDLDVPEMPSIPNPFPELCCSPITSVLSADLFPKANSRKKQVIKVYSEDETSRALDV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name GRB14
Gene Symbol GRB14
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.