Learn More
Description
This product is a solid phase sandwich ELISA for the measurement of natural and recombinant human Gremlin.
- Assay Type: Solid phase sandwich ELISA
- Detection Method: Colorimetric (450 nm)
- Sample Types: Cell culture supernates, serum, and plasma
- Assay Range: 125 to 4000 pg/mL
- Reactivity: Human
- Gene Symbol: GREM1
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 26585 |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Signaling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
