Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GRK3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
| Antigen | GRK3 |
|---|---|
| Dilution | Western Blot 1 ug/mL, Simple Western 20 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GRK3 Polyclonal specifically detects GRK3 in Human samples. It is validated for Western Blot, Simple Western.Specifications
| GRK3 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRBK2, adrenergic, beta, receptor kinase 2, BARK2, BARK2beta-adrenergic receptor kinase 2, beta-ARK-2, EC 2.7.11, EC 2.7.11.15, G-protein-coupled receptor kinase 3, GRK3Beta-ARK-2 | |
| E. coli-derived recombinant human GRK3 Pro469-Leu688 Accession # P35626 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human GRK3 in direct ELISAs and Western blots. |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| P35626 | |
| 157 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title