Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Growth Hormone Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.50
Specifications
Antigen | GrowthHormone |
---|---|
Clone | 178915R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10671SP
![]() |
R&D Systems
MAB10671SP |
25 μg |
Each for $214.00
|
|
|||||
MAB10671100
![]() |
R&D Systems
MAB10671100 |
100 μg |
Each for $561.50
|
|
|||||
Description
Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for ELISA.Specifications
GrowthHormone | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
CAA23779 | |
2688 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
178915R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
E. coli-derived recombinant human Growth Hormone Phe27-Phe217 Accession # CAA23779 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Growth Hormone in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title