Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Growth Hormone R/GHR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1210SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1210SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1210SP Supplier R&D Systems Supplier No. MAB1210SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Growth Hormone R Monoclonal specifically detects Growth Hormone R in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Growth Hormone R
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 193804
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.025-0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10912
Gene Alias GH receptor, GHBP, GHR, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor
Gene Symbols GHR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Growth Hormone R/GHR Phe19-Tyr264 Accession # P10912
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2690
Test Specificity Detects human Growth Hormone R/GHR in Western blots and direct ELISAs. In Western blots, no cross-reactivity with recombinant mouse GHR or recombinant rat GHR is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.