Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GRP75/HSPA9B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB35841SP
This item is not returnable.
View return policy
Description
GRP75/HSPA9B/Mortalin Monoclonal specifically detects GRP75/HSPA9B/Mortalin in Human samples. It is validated for Immunohistochemistry.Specifications
GRP75/HSPA9B/Mortalin | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P38646 | |
HSPA9 | |
E. coli-derived recombinant human GRP75/HSPA9B Glu542-Gln679 Accession # P38646 | |
25 μg | |
Primary | |
Detects human GRP75/HSPA9B in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
419605 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
75 kDa glucose-regulated protein, CSA, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9, HSPA9B, HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3313 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction