Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GRP75/HSPA9B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB35841
This item is not returnable.
View return policy
Description
GRP75/HSPA9B/Mortalin Monoclonal specifically detects GRP75/HSPA9B/Mortalin in Human samples. It is validated for Immunohistochemistry.Specifications
GRP75/HSPA9B/Mortalin | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P38646 | |
HSPA9 | |
E. coli-derived recombinant human GRP75/HSPA9B Glu542-Gln679 Accession # P38646 | |
100 μg | |
Primary | |
Detects human GRP75/HSPA9B in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
419605 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
75 kDa glucose-regulated protein, CSA, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9, HSPA9B, HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3313 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction