Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Guanylyl Cyclase C/GUCY2C Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2157S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2157S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2157S100 Supplier R&D Systems Supplier No. FAB2157S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GUCY2C Monoclonal specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GUCY2C
Applications Flow Cytometry
Classification Monoclonal
Clone 2543C
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P25092
Gene Alias DIAR6, EC 4.6.1, GCC, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylate Cyclase C, Guanylyl cyclase C, GUC2C, GUC2CEC 4.6.1.2, GUCY2C, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, MUCIL, STA Receptor, StAR, STARSTA receptor
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived human Guanylyl Cyclase C/GUCY2C, Ser21-Gln430, Accession # P25092
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2984
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.