Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human H2BFWT Partial ORF (NP_001002916.1, 31 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89946983
Click to view available options
Quantity:
2 μg

Used for AP, Array, ELISA, WB-Re

Testis-specific histones, like H2BFWT, are synthesized and accumulate at specific stages of mammalian spermatogenesis. Their proposed functions range from facilitation of the replacement of somatic histones by protamines to epigenetic control of gene transcription (Churikov et al., 2004 [PubMed 15475252]).[supplied by OMIM]

Sequence: GPSSETTSEEQLITQEPKEANSTTSQKQSKQRKRGRHGPRRCHSNCRGDSFATYFRRVLKQVHQGLSLSREAVSVMDSLVHDILDRIATEAGHLARSTKR

Specifications

Accession Number NP_001002916.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 158983
Molecular Weight (g/mol) 36.74kDa
Name H2BFWT (Human) Recombinant Protein (Q01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen GPSSETTSEEQLITQEPKEANSTTSQKQSKQRKRGRHGPRRCHSNCRGDSFATYFRRVLKQVHQGLSLSREAVSVMDSLVHDILDRIATEAGHLARSTKR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC148130/MGC148131
Common Name H2BFWT
Gene Symbol H2BFWT
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.