Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HAI-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1048
This item is not returnable.
View return policy
Description
HAI-1/HGFA Inhibitor 1 Monoclonal specifically detects HAI-1/HGFA Inhibitor 1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HAI-1/HGFA Inhibitor 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
O43278 | |
SPINT1 | |
Mouse myeloma cell line NS0-derived recombinant human HAI-1 Pro37-Val449 Accession # O43278 | |
500 μg | |
Primary | |
Detects human HAI-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human HAI-2 or recombinant mouse HAI-1 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
169417 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1, SPINT1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6692 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction