Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HAI-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1048SP
This item is not returnable.
View return policy
Description
HAI-1/HGFA Inhibitor 1 Monoclonal specifically detects HAI-1/HGFA Inhibitor 1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HAI-1/HGFA Inhibitor 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
O43278 | |
SPINT1 | |
Mouse myeloma cell line NS0-derived recombinant human HAI-1 Pro37-Val449 Accession # O43278 | |
25 μg | |
Primary | |
Detects human HAI-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human HAI-2 or recombinant mouse HAI-1 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
169417 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1, SPINT1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6692 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction