Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human HAI-1 DuoSet ELISA
SDP

Catalog No. DY1048 Shop All Novus Biologicals Products
Change view
Click to view available options
Conjugate:
Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
DY1048 Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY1048 Supplier R&D Systems™ Supplier No. DY1048

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Human HAI-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human HAI-1/HGFA Inhibitor 1.

Specifications

Product Type Solid Phase Sandwich ELISA Development Kit
Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Range 156.00 to 10000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 6692
Product Line R&D Systems
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.