Learn More
Novus Biologicals™ Human HAI-1/HGFA Inhibitor 1 ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 0.8 to 200 ng/mL |
| Assay Sensitivity | 0.8 ng/ml |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Specificity | This ELISA antibody pair detects human HAI-1/HGFA Inhibitor 1, no cross-reactivity with mouse HAI-1/HGFA Inhibitor 1, human APP, GASP-1, GASP-2, HAI-2A, TFPI, TFPI-2 is observed. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Gene ID (Entrez) | 6692 |
| Gene Symbol | SPINT1 |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.