Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Hamster ACE-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB933
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB933 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB933 Supplier R&D Systems Supplier No. MAB933
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACE-2 Monoclonal specifically detects ACE-2 in Human, Hamster samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ACE-2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 171606
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BYF1
Gene Alias ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Gene Symbols ACE2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ACE-2 Gln18-Ser740 (predicted) Accession # Q9BYF1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59272
Test Specificity Detects human ACE-2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ACE-1 or rhNeprilysin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Hamster
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.