Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HAPLN1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2608
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2608 100 μg
MAB2608SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2608 Supplier R&D Systems Supplier No. MAB2608
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HAPLN1 Monoclonal specifically detects HAPLN1 in Human samples. It is validated for Western Blot.

Specifications

Antigen HAPLN1
Applications Western Blot
Classification Monoclonal
Clone 316119
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10915
Gene Alias Cartilage link protein, Cartilage-link protein, Cartilage-linking protein 1, CRTL1, CRTL1cartilage linking protein 1, hyaluronan and proteoglycan link protein 1, Proteoglycan link protein
Gene Symbols HAPLN1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human HAPLN1 Asp16-Asn354 Accession # P10915
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1404
Test Specificity Detects human HAPLN1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human HAPLN4.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.