Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HAPLN1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2608
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2608 100 μg
AF2608SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2608 Supplier R&D Systems Supplier No. AF2608
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

HAPLN1 Polyclonal specifically detects HAPLN1 in Human samples. It is validated for Western Blot.

Specifications

Antigen HAPLN1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P10915
Gene Alias Cartilage link protein, Cartilage-link protein, Cartilage-linking protein 1, CRTL1, CRTL1cartilage linking protein 1, hyaluronan and proteoglycan link protein 1, Proteoglycan link protein
Gene Symbols HAPLN1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human HAPLN1 (R&D Systems, Catalog # 2608-HP) Asp16-Asn354 Accession # P10915
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1404
Test Specificity Detects human HAPLN1 in direct ELISAs and Western blots. In these formats, this antibody shows approximately 5% cross-reactivity with recombinant human HAPLN4.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.