Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HAPLN4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4085SP
This item is not returnable.
View return policy
Description
HAPLN4 Polyclonal specifically detects HAPLN4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
HAPLN4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BRAL2, BRAL2KIAA1926Brain link protein 2, hyaluronan and proteoglycan link protein 4, Lpr4 | |
Goat | |
Affinity Purified | |
RUO | |
404037 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Q86UW8 | |
HAPLN4 | |
Mouse myeloma cell line NS0-derived recombinant human HAPLN4 Gln30-Val402 Accession # Q86UW8 | |
25 μg | |
Primary | |
Detects human HAPLN4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human HAPLN1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction