Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HAX-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5458SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5458SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5458SP Supplier R&D Systems Supplier No. AF5458SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

HAX-1 Polyclonal specifically detects HAX-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen HAX-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00165
Gene Alias FLJ17042, FLJ93803, HAX1, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1, HS1BP1FLJ18492, HSP1BP-1, SCN3
Gene Symbols HAX1
Host Species Goat
Immunogen E. coli-derived recombinant human HAX-1 Ser2-Leu261 Accession # O00165
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10456
Test Specificity Detects human HAX-1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.