Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HB-EGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB259SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB259SP 25 μg
MAB259100 100 μg
MAB259500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB259SP Supplier R&D Systems Supplier No. MAB259SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot.

Specifications

Antigen HB-EGF
Applications Western Blot
Classification Monoclonal
Clone 125923
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q53H93
Gene Alias diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
Gene Symbols HBEGF
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF Asp63-Leu148 Accession # Q53H93
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1839
Test Specificity Detects human HB-EGF in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse HB-EGF. Does not cross-react with recombinant human (rh) AR, rhBTC, rhHRG-alpha , rhHRG-beta , rhEGF, or rhTGF-alpha .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.