Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HB-EGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $2020.00
Specifications
Antigen | HB-EGF |
---|---|
Clone | 406316 |
Dilution | Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2591SP
![]() |
R&D Systems
MAB2591SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2591100
![]() |
R&D Systems
MAB2591100 |
100 μg |
Each for $549.50
|
|
|||||
MAB2591500
![]() |
R&D Systems
MAB2591500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Neutralization.Specifications
HB-EGF | |
Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
Q53H93 | |
1839 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF Asp63-Leu148 Accession # Q53H93 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human HB-EGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Amphiregulin, rhBTC, rhHRG-alpha , rhHRG-beta , rhEGF, rhTGF-alpha , recombinant mouse (rm) Epigen, or rmEpiregulin. |
406316 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
HBEGF | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title