Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HB-EGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF259NA
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF259NA 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF259NA Supplier R&D Systems Supplier No. AF259NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

HB-EGF Polyclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen HB-EGF
Applications Western Blot, Immunohistochemistry, Neutralization, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. Q99075
Gene Alias diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
Gene Symbols HBEGF
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF (R&D Systems, Catalog # 259-HE) Asp63-Leu148 Accession # Q99075
Purification Method Affinity Purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1839
Test Specificity Detects human HB-EGF in ELISAs and Western blots. In direct ELISAs, less than 1% cross reactivity with recombinant mouse HB-EGF is observed. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) Amphiregulin, rhBetacellulin, rhEpiregulin, and recombinant mouse Epigen is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.