Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human HB-EGF Recombinant Protein, PeproTech®

Catalog No. p-7245962
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human HB-EGF is a 9.7 kDa protein containing 86 amino acid residues, corresponding to the extracellular EGF-like and heparin-binding domains of the full length HB-EGF protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

HBEGF is a growth factor that mediates its effects via EGFR, ERBB2 and ERBB4. It is required for normal cardiac valve formation and normal heart function. HBEGF promotes smooth muscle cell proliferation. It may be involved in macrophage-mediated cellular proliferation. It is mitogenic for fibroblasts, but not endothelial cells. It is able to bind EGF receptor/EGFR with higher affinity than EGF itself and is a far more potent mitogen for smooth muscle cells than EGF. It also acts as a diphtheria toxin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99075
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 1839
Molecular Weight (g/mol) 9.7 kDa
Name Human HB-EGF
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AW047313; Diphtheria toxin receptor; Diphtheria toxin receptor (heparin binding epidermal growth factor - like growth factor); diphtheria toxin receptor (heparin-binding EGF-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor); DTR; DT-R; DTS; DTSF; GFHB; hb egf; HBEGF; Hb-egf; Hegfl; heparin binding EGF like growth factor; heparin binding epidermal growth factor-like growth factor; heparin-binding EGF-like growth factor; heparin-binding epidermal -growth - like growth factor; heparin-binding epidermal growth factor; heparin-binding epidermal -growth factor; proheparin-binding EGF-like growth factor
Common Name HBEGF
Gene Symbol HBEGF
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence DLQEADLDLL RVTLSSKPQA LATPNKEEHG KRKKKGKGLG KKRDPCLRKY KDFCIHGECK YVKELRAPSC ICHPGYHGER CHGLSL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.