Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HCN4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8138
This item is not returnable.
View return policy
Description
HCN4 Polyclonal specifically detects HCN4 in Human samples. It is validated for Immunocytochemistry.Specifications
HCN4 | |
Polyclonal | |
Unconjugated | |
Q9Y3Q4 | |
HCN4 | |
E. coli-derived recombinant human HCN4 Lys1084-Leu1203 Accession # Q9Y3Q4 | |
100 μg | |
Primary | |
Detects human HCN4 in direct ELISAs. | |
Human | |
IgG |
Immunocytochemistry | |
LYOPH | |
Immunocytochemistry 5-15 ug/mL | |
hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2 | |
Sheep | |
Affinity Purified | |
RUO | |
10021 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction