Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HE4/WFDC2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | HE4/WFDC2 |
---|---|
Clone | 676009 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB62741SP
![]() |
R&D Systems
MAB62741SP |
25 μg |
Each for $200.50
|
|
|||||
MAB62741100
![]() |
R&D Systems
MAB62741100 |
100 μg |
Each for $514.50
|
|
|||||
Description
HE4/WFDC2 Monoclonal specifically detects HE4/WFDC2 in Human samples. It is validated for ELISA.Specifications
HE4/WFDC2 | |
ELISA | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5, WAP5epididymal protein 4, WFDC2 | |
Mouse myeloma cell line NS0-derived recombinant human HE4/WFDC2, Thr28-Phe124, Accession No. Q14508 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human HE4/WFDC2 in direct ELISAs. |
676009 | |
ELISA | |
Unconjugated | |
Human | |
Q14508 | |
10406.0 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title