Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HE4/WFDC2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6274
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6274 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6274 Supplier R&D Systems Supplier No. AF6274
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

HE4/WFDC2 Polyclonal specifically detects HE4/WFDC2 in Human samples. It is validated for ELISA, Immunoprecipitation.

Specifications

Antigen HE4/WFDC2
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA, Immunoprecipitation 25 ug/mL
Gene Accession No. Q14508
Gene Alias dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5, WAP5epididymal protein 4, WFDC2
Gene Symbols WFDC2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human HE4/WFDC2 Thr28-Phe124 Accession # Q14508
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10406
Test Specificity Detects human HE4/WFDC2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.