Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Helios Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC73092U100UG
This item is not returnable.
View return policy
Description
Helios Monoclonal specifically detects Helios in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Helios | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UKS7 | |
IKZF2 | |
E. coli-derived recombinant human Helios Met1-Gln97 Accession # Q9UKS7 | |
RUO | |
22807 | |
Human | |
IgG1 |
Flow Cytometry | |
736440 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, IKZF2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
Mouse | |
100 μg | |
Primary | |
Detects human Helios in direct ELISA. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction