Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Helios Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB73091
This item is not returnable.
View return policy
Description
Helios Monoclonal specifically detects Helios in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Helios | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
Q9UKS7 | |
IKZF2 | |
E. coli-derived recombinant human Helios Thr3-Gln97 Accession # Q9UKS7 | |
100 μg | |
Primary | |
Detects human Helios in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
736423 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, IKZF2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22807 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction