Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Hemoglobin zeta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7708SP
This item is not returnable.
View return policy
Description
Hemoglobin zeta Monoclonal specifically detects Hemoglobin zeta in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Hemoglobin zeta | |
Monoclonal | |
LYOPH | |
Western Blot 0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P02008 | |
HBZ | |
E. coli-derived recombinant human Hemoglobin zeta Met1-Arg142 Accession # P02008 | |
25 μg | |
Primary | |
Detects human Hemoglobin zeta in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
778633 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HBAZ, HBZ, HBZ2, hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, zeta, zeta-globin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3050 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction