Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5968SP
This item is not returnable.
View return policy
Description
Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 Polyclonal specifically detects Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3OST, 3OST1, 3-OST-1, 3OST1Heparan sulfate 3-O-sulfotransferase 1, EC 2.8.2, EC 2.8.2.23, h3-OST-1, heparan sulfate (glucosamine) 3-O-sulfotransferase 1, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1, heparan sulfate glucosamine 3-O-sulfotransferase 1, heparin-glucosamine 3-O-sulfotransferase | |
Sheep | |
Affinity Purified | |
RUO | |
9957 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
O14792 | |
HS3ST1 | |
Chinese hamster ovary cell line CHO-derived recombinant human HS3ST1 Arg21-His307 Accession # O14792 | |
25 μg | |
Primary | |
Detects human HS3ST1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction